Learn More
Cayman Chemical Myristic Acid methyl ester, Purity: ≥95%, CAS: 124-10-7, 1 g
SDP
Catalog No. 502895072
Encompass_Preferred
An esterified version of the free acid, which is less water soluble but more amenable for the formulation of myristate-containing diets and dietary supplements
Catalog No. 50-289-5072 Supplier Cayman Chemical Supplier No. 90018671 G
An esterified version of the free acid, which is less water soluble but more amenable for the formulation of myristate-containing diets and dietary supplements