Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myristic Anhydride 95.0+%, TCI America™
SDP

Catalog No. M048425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M048425G 25 g
M0484250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M048425G Supplier TCI America Supplier No. M048425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 626-29-9
Molecular Formula C28H54O3
Molecular Weight (g/mol) 438.737
MDL Number MFCD00056151
InChI Key RCRYHUPTBJZEQS-UHFFFAOYSA-N
PubChem CID 69377
IUPAC Name tetradecanoyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 54°C
Color White
Quantity 25 g
Formula Weight 438.74
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Myristic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.