Learn More
Medchemexpress LLC Myristic-d27 acid | 60658-41-5 | MFCD00053329 | 255.54 | 25 MG
SDP
Catalog No. 5000389309
Encompass_Preferred
Myristic acid-d27 is the deuterium labeled Myristic acid. Myristic acid is a saturated 14-carbon fatty acid occurring in most animal and vegetable fats, particularly butterfat and coconut, palm, and nutmeg oils. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Saturated 14-carbon fatty acid
  • Occurs in most animal and vegetable fats
Catalog No. 50-003-89309 Supplier Medchemexpress LLC Supplier No. 25MGHYN2041S
Myristic acid-d27 is the deuterium labeled Myristic acid. Myristic acid is a saturated 14-carbon fatty acid occurring in most animal and vegetable fats, particularly butterfat and coconut, palm, and nutmeg oils. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Saturated 14-carbon fatty acid
  • Occurs in most animal and vegetable fats