Learn More
Selleck Chemical LLC Myristicin S3291-5mg
SDP
Catalog No. 503109832
Encompass_Preferred
Myristicin (Myristicine) a terpene-like active component in Nutmeg which is the seed of Myristica fragrans is a weak inhibitor of monamine oxidase (MAO) and a mechanism-based inhibitor of CYP1A2
Catalog No. 50-310-9832 Supplier Selleck Chemical LLC Supplier No. S32915MG
Myristicin (Myristicine) a terpene-like active component in Nutmeg which is the seed of Myristica fragrans is a weak inhibitor of monamine oxidase (MAO) and a mechanism-based inhibitor of CYP1A2