Learn More
Medchemexpress LLC Myristicin | 607-91-0 | 100 MG
SDP
Catalog No. 5000421123
Encompass_Preferred
Myristicin is an orally bioavailable serotonin receptor antagonist and a weak monoamine oxidase (MAO) inhibitor. It also exhibits anti-cancer effects on gastric cancer cells by inhibiting the EGFR/ERK signaling pathway. Myristicin is the main component of nutmeg essential oil and possesses anti-cancer, anti-proliferative, antibacterial, anti-inflammatory, and apoptosis-inducing properties.
  • Serotonin receptor antagonist
  • Weak monoamine oxidase (MAO) inhibitor
  • Inhibits EGFR/ERK signaling pathway in gastric cancer cells
  • Anti-cancer and anti-proliferative effects
  • Antibacterial and anti-inflammatory properties
  • Induces apoptosis
  • Main component of nutmeg essential oil
Catalog No. 50-004-21123 Supplier Medchemexpress LLC Supplier No. HYN2510100MG
Myristicin is an orally bioavailable serotonin receptor antagonist and a weak monoamine oxidase (MAO) inhibitor. It also exhibits anti-cancer effects on gastric cancer cells by inhibiting the EGFR/ERK signaling pathway. Myristicin is the main component of nutmeg essential oil and possesses anti-cancer, anti-proliferative, antibacterial, anti-inflammatory, and apoptosis-inducing properties.
  • Serotonin receptor antagonist
  • Weak monoamine oxidase (MAO) inhibitor
  • Inhibits EGFR/ERK signaling pathway in gastric cancer cells
  • Anti-cancer and anti-proliferative effects
  • Antibacterial and anti-inflammatory properties
  • Induces apoptosis
  • Main component of nutmeg essential oil