Learn More
Medchemexpress LLC Myristicin | 607-91-0 | 25 MG
SDP
Catalog No. 5000422065
Encompass_Preferred
Myristicin is an orally bioavailable serotonin receptor antagonist and a weak monoamine oxidase (MAO) inhibitor. It exhibits anti-cancer effects on gastric cancer cells by inhibiting the EGFR/ERK signaling pathway. As the primary component of nutmeg essential oil, Myristicin possesses anti-cancer, anti-proliferative, antibacterial, anti-inflammatory, and apoptosis-inducing properties.
  • Orally bioavailable serotonin receptor antagonist
  • Weak monoamine oxidase (MAO) inhibitor
  • Inhibits EGFR/ERK signaling pathway
  • Main component of nutmeg essential oil
  • Exhibits anti-cancer, anti-proliferative, antibacterial, and anti-inflammatory properties
  • Induces apoptosis in gastric cancer cells
  • Reduces cell cycle protein expression
  • Increases Bax expression and activates caspase enzyme
  • Targets EGFR, ERK, and 5-HT Receptor
Catalog No. 50-004-22065 Supplier Medchemexpress LLC Supplier No. 25MGHYN2510
Myristicin is an orally bioavailable serotonin receptor antagonist and a weak monoamine oxidase (MAO) inhibitor. It exhibits anti-cancer effects on gastric cancer cells by inhibiting the EGFR/ERK signaling pathway. As the primary component of nutmeg essential oil, Myristicin possesses anti-cancer, anti-proliferative, antibacterial, anti-inflammatory, and apoptosis-inducing properties.
  • Orally bioavailable serotonin receptor antagonist
  • Weak monoamine oxidase (MAO) inhibitor
  • Inhibits EGFR/ERK signaling pathway
  • Main component of nutmeg essential oil
  • Exhibits anti-cancer, anti-proliferative, antibacterial, and anti-inflammatory properties
  • Induces apoptosis in gastric cancer cells
  • Reduces cell cycle protein expression
  • Increases Bax expression and activates caspase enzyme
  • Targets EGFR, ERK, and 5-HT Receptor