Learn More
Medchemexpress LLC Myristicin | 607-91-0 | 50 MG
SDP
Catalog No. 5000421326
Encompass_Preferred
Myristicin is an orally bioavailable serotonin receptor antagonist and a weak monoamine oxidase (MAO) inhibitor. It is also the main component of nutmeg essential oil and demonstrates anti-cancer effects on gastric cancer cells by inhibiting the EGFR/ERK signaling pathway.
  • Possesses anti-cancer, anti-proliferative, antibacterial, anti-inflammatory, and apoptosis-inducing properties.
  • Delays the proliferation of gastric cancer cells.
  • Induces endoplasmic reticulum (ER) stress and apoptosis.
  • Reduces cell cycle protein expression.
  • Increases Bax expression.
  • Activates caspase enzyme.
  • Enhances cytochrome C release and mitochondrial ROS levels.
  • Inhibits the growth of gastric cancer cells and the EGFR/ERK signaling pathway in animal models.
  • Has preventive and protective effects against acetic acid-induced experimental colitis.
Catalog No. 50-004-21326 Supplier Medchemexpress LLC Supplier No. HYN251050MG
Myristicin is an orally bioavailable serotonin receptor antagonist and a weak monoamine oxidase (MAO) inhibitor. It is also the main component of nutmeg essential oil and demonstrates anti-cancer effects on gastric cancer cells by inhibiting the EGFR/ERK signaling pathway.
  • Possesses anti-cancer, anti-proliferative, antibacterial, anti-inflammatory, and apoptosis-inducing properties.
  • Delays the proliferation of gastric cancer cells.
  • Induces endoplasmic reticulum (ER) stress and apoptosis.
  • Reduces cell cycle protein expression.
  • Increases Bax expression.
  • Activates caspase enzyme.
  • Enhances cytochrome C release and mitochondrial ROS levels.
  • Inhibits the growth of gastric cancer cells and the EGFR/ERK signaling pathway in animal models.
  • Has preventive and protective effects against acetic acid-induced experimental colitis.