Learn More
Cayman Chemical Myristoleic Acid, Purity: ≥95%, CAS: 544-64-9, 100 mg
SDP
Catalog No. 502894168
Encompass_Preferred
A bioactive fatty acid; induces apoptosis and necrosis in human prostate cancer LNCaP cells at a rate of 8.8% and 8.1%, respectively; inhibits C. albicans germination (MIC = 9 µM); inhibits osteogenesis in vitro via interference with cytoskeletal rearrangement; prevents RANKL-induced bone loss and osteoclast formation in mice
Catalog No. 50-289-4168 Supplier Cayman Chemical Supplier No. 9002461100 MG
A bioactive fatty acid; induces apoptosis and necrosis in human prostate cancer LNCaP cells at a rate of 8.8% and 8.1%, respectively; inhibits C. albicans germination (MIC = 9 µM); inhibits osteogenesis in vitro via interference with cytoskeletal rearrangement; prevents RANKL-induced bone loss and osteoclast formation in mice