Learn More
Medchemexpress LLC Myristoleic acid | 544-64-9 | 98.0% | 226.36 | 100 MG
SDP
Catalog No. 5000435917
Encompass_Preferred
Myristoleic acid is a cytotoxic component found in the extract of Serenoa repens. It induces apoptosis and necrosis in human prostatic LNCaP cells. It also inhibits RANKL-induced osteoclast formation in vitro, particularly at later stages of differentiation, and has been shown to prevent RANKL-induced bone loss and osteoclast formation in mice.
  • Cytotoxic component from *Serenoa repens*.
  • Induces apoptosis and necrosis in LNCaP cells.
  • Inhibits RANKL-induced osteoclast formation in vitro.
  • Prevents RANKL-induced bone loss in mice.
  • For research use only.
Catalog No. 50-004-35917 Supplier Medchemexpress LLC Supplier No. HY113332100MG
Myristoleic acid is a cytotoxic component found in the extract of Serenoa repens. It induces apoptosis and necrosis in human prostatic LNCaP cells. It also inhibits RANKL-induced osteoclast formation in vitro, particularly at later stages of differentiation, and has been shown to prevent RANKL-induced bone loss and osteoclast formation in mice.
  • Cytotoxic component from *Serenoa repens*.
  • Induces apoptosis and necrosis in LNCaP cells.
  • Inhibits RANKL-induced osteoclast formation in vitro.
  • Prevents RANKL-induced bone loss in mice.
  • For research use only.