Learn More
Cayman Chemical Myristoleic Acid methyl ester, Purity: ≥98%, CAS: 56219-06-8, 1 g
SDP
Catalog No. 502888962
Encompass_Preferred
A more hydrophobic form of the free acid, a cytotoxic component from the fruit extract of S. repens
Catalog No. 50-288-8962 Supplier Cayman Chemical Supplier No. 100085811 G
A more hydrophobic form of the free acid, a cytotoxic component from the fruit extract of S. repens