Learn More
Cayman Chemical Myristoleic Acid methyl ester, Purity: ≥98%, CAS: 56219-06-8, 100 mg
SDP
Catalog No. 502888963
Encompass_Preferred
A more hydrophobic form of the free acid, a cytotoxic component from the fruit extract of S. repens
Catalog No. 50-288-8963 Supplier Cayman Chemical Supplier No. 10008581100 MG
A more hydrophobic form of the free acid, a cytotoxic component from the fruit extract of S. repens