Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myristonitrile 96.0+%, TCI America™
SDP

Catalog No. M0740500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0740500ML 500 mL
M074025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0740500ML Supplier TCI America Supplier No. M0740500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 629-63-0
Molecular Formula C14H27N
Molecular Weight (g/mol) 209.38
MDL Number MFCD00019848
InChI Key MLRCLPRHEOPXLL-UHFFFAOYSA-N
Synonym myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3
PubChem CID 64678
IUPAC Name tetradecanenitrile
SMILES CCCCCCCCCCCCCC#N

Specifications

Color Yellow
Boiling Point 227°C
Quantity 500 mL
Formula Weight 209.38
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Myristonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.