Learn More
Medchemexpress LLC Myristoyl coenzyme A triammonium | 799812-85-4 | 99% | 1 MG
SDP
Catalog No. 5000342493
Encompass_Preferred
Myristoyl coenzyme A triammonium (14:0 Coenzyme A) is a long-chain acyl-CoA, which is the activated form of long-chain fatty acids and serves as a crucial lipid metabolite.
  • Long-chain acyl-CoA
  • Activated form of long-chain fatty acids
  • Crucial lipid metabolite
Catalog No. 50-003-42493 Supplier Medchemexpress LLC Supplier No. HY126833B1MG
Myristoyl coenzyme A triammonium (14:0 Coenzyme A) is a long-chain acyl-CoA, which is the activated form of long-chain fatty acids and serves as a crucial lipid metabolite.
  • Long-chain acyl-CoA
  • Activated form of long-chain fatty acids
  • Crucial lipid metabolite