Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myristoyl glutamic acid sodium | 38517-37-2 | 98.43% | 5 G
SDP

Catalog No. 5000339157
Encompass_Preferred

Myristoyl glutamic acid sodium is a common ingredient in cosmetics and creams. This surfactant belongs to the amino acid class, possessing mild cleaning properties, and is also foamy and mildly bacteriostatic.

  • Molecular weight: 379.47
  • Formula: C19H34NNaO5
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-39157 Supplier Medchemexpress LLC Supplier No. HY126460A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myristoyl glutamic acid sodium is a common ingredient in cosmetics and creams. This surfactant belongs to the amino acid class, possessing mild cleaning properties, and is also foamy and mildly bacteriostatic.

  • Molecular weight: 379.47
  • Formula: C19H34NNaO5
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.