Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myristoyl-L-carnitine chloride | 173686-73-2 | MFCD00274133 | 99.9% | 408.02 g/mol | C21H42ClNO4 | 5 MG
SDP

Catalog No. 5000223402
Encompass_Preferred

Myristoyl-L-carnitine chloride is the myristoyl ester of L-carnitine supplied as the chloride salt. It is a long-chain acylcarnitine commonly used as a research reagent in studies of fatty acid metabolism and acylcarnitine biology. The compound is supplied as a solid with reported high purity and should be stored sealed at low temperature to maintain stability.

  • Long-chain acylcarnitine chloride salt suitable for metabolic studies.
  • High purity: 99.9% (as reported).
  • Solid form, easy to weigh and handle.
  • Molecular weight 408.02 g/mol and formula C21H42ClNO4.
  • Stable when stored sealed at 4°C and protected from moisture.
  • Available in small research pack sizes for laboratory use.

Catalog No. 50-002-23402 Supplier Medchemexpress LLC Supplier No. HY1351705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Myristoyl-L-carnitine chloride is the myristoyl ester of L-carnitine supplied as the chloride salt. It is a long-chain acylcarnitine commonly used as a research reagent in studies of fatty acid metabolism and acylcarnitine biology. The compound is supplied as a solid with reported high purity and should be stored sealed at low temperature to maintain stability.

  • Long-chain acylcarnitine chloride salt suitable for metabolic studies.
  • High purity: 99.9% (as reported).
  • Solid form, easy to weigh and handle.
  • Molecular weight 408.02 g/mol and formula C21H42ClNO4.
  • Stable when stored sealed at 4°C and protected from moisture.
  • Available in small research pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.