Learn More
Medchemexpress LLC Myristoyl-L-carnitine-d3 (chloride) | 1334532-25-0 | MFCD08705130; MFCD32063383 | 99.9% | 411.03 | C21H39D3ClNO4 | 5 MG
SDP
Catalog No. 5000222510
Encompass_Preferred
Myristoyl-L-carnitine-d3 (chloride) is a deuterium-labeled analytical standard of myristoyl-L-carnitine supplied as the chloride salt. It is intended for research use as an internal standard in LC-MS/MS and metabolic profiling workflows, providing reliable quantitation in analytical assays. Supplied in small quantities for laboratory analytical applications.
  • Deuterium-labeled (d3) isotopologue for accurate mass spectrometry quantitation.
  • Chloride salt form for stability and handling.
  • High purity (99.9% by HPLC) for analytical applications.
  • Supplied in a 5 mg package suitable for method development and standards use.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-22510 Supplier Medchemexpress LLC Supplier No. HY135170S5MG
Myristoyl-L-carnitine-d3 (chloride) is a deuterium-labeled analytical standard of myristoyl-L-carnitine supplied as the chloride salt. It is intended for research use as an internal standard in LC-MS/MS and metabolic profiling workflows, providing reliable quantitation in analytical assays. Supplied in small quantities for laboratory analytical applications.
  • Deuterium-labeled (d3) isotopologue for accurate mass spectrometry quantitation.
  • Chloride salt form for stability and handling.
  • High purity (99.9% by HPLC) for analytical applications.
  • Supplied in a 5 mg package suitable for method development and standards use.
  • For research use only; not for human or clinical applications.