Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myristoyl-(Lys12,27,28)-VIP-Gly-Gly-Thr free acid | 2243219-86-3 | 99.71% | 3753.42 | 1 MG
SDP

Catalog No. 5000399949
Encompass_Preferred

Myristoyl-(Lys12,27,28)-VIP-Gly-Gly-Thr (free acid) is a high-affinity and selective VPAC2 receptor antagonist. It is characterized by its high purity and specific molecular weight.

  • High-affinity VPAC2 receptor antagonist
  • Selective VPAC2 receptor antagonist
  • Purity of 99.71%
  • Molecular weight of 3753.42

Catalog No. 50-003-99949 Supplier Medchemexpress LLC Supplier No. HYP51111MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myristoyl-(Lys12,27,28)-VIP-Gly-Gly-Thr (free acid) is a high-affinity and selective VPAC2 receptor antagonist. It is characterized by its high purity and specific molecular weight.

  • High-affinity VPAC2 receptor antagonist
  • Selective VPAC2 receptor antagonist
  • Purity of 99.71%
  • Molecular weight of 3753.42

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.