Learn More
Medchemexpress LLC Myristoyl pentapeptide-4 | 1392416-25-9 | 98.9% | 774.00 | 1 MG
SDP
Catalog No. 5000398951
Encompass_Preferred
Myristoyl pentapeptide-4 is a bioactive peptide used as a cosmetic ingredient to promote hair growth. This synthetic peptide, composed of Lys-Thr-Thr-Lys-Ser (KTTKS), targets keratin to activate gene transcription and translation, enhancing hair elasticity and strength. It also stimulates HaCaT cell proliferation, human hair follicle papilla cell migration, and skin fibroblasts to produce collagen and fibronectin, aiding in wrinkle reduction, improved skin elasticity, wound healing, and skin repair.
  • Targets keratin to activate gene transcription and translation
  • Enhances hair elasticity and strength
  • Promotes HaCaT cell proliferation
  • Stimulates human hair follicle papilla cell migration
  • Stimulates skin fibroblasts to produce collagen and fibronectin
  • Aids in wrinkle reduction and improved skin elasticity
  • Promotes wound healing and skin repair
Catalog No. 50-003-98951 Supplier Medchemexpress LLC Supplier No. HYP52391MG
Myristoyl pentapeptide-4 is a bioactive peptide used as a cosmetic ingredient to promote hair growth. This synthetic peptide, composed of Lys-Thr-Thr-Lys-Ser (KTTKS), targets keratin to activate gene transcription and translation, enhancing hair elasticity and strength. It also stimulates HaCaT cell proliferation, human hair follicle papilla cell migration, and skin fibroblasts to produce collagen and fibronectin, aiding in wrinkle reduction, improved skin elasticity, wound healing, and skin repair.
  • Targets keratin to activate gene transcription and translation
  • Enhances hair elasticity and strength
  • Promotes HaCaT cell proliferation
  • Stimulates human hair follicle papilla cell migration
  • Stimulates skin fibroblasts to produce collagen and fibronectin
  • Aids in wrinkle reduction and improved skin elasticity
  • Promotes wound healing and skin repair