Learn More
Cayman Chemical Myristoyl Pentapeptide-17 (trifluoroacetate salt), Purity: ≥95%, 25 mg
SDP
Catalog No. 502888332
Encompass_Preferred
A myristoylated form of the pentapeptide Lys-Leu-Ala-Lys-Lys-NH2; it is active against E. coli (MIC = 10 ppm)
Catalog No. 50-288-8332 Supplier Cayman Chemical Supplier No. 1052625 MG
A myristoylated form of the pentapeptide Lys-Leu-Ala-Lys-Lys-NH2; it is active against E. coli (MIC = 10 ppm)