Learn More
Cayman Chemical Myristoyl Pentapeptide17 ace
SDP
Catalog No. 502891114
Encompass_Preferred
A myristoylated form of the pentapeptide Lys-Leu-Ala-Lys-Lys-NH2; it is active against E. coli (MIC = 10 ppm)
Catalog No. 50-289-1114 Supplier Cayman Chemical Supplier No. 105265 MG
A myristoylated form of the pentapeptide Lys-Leu-Ala-Lys-Lys-NH2; it is active against E. coli (MIC = 10 ppm)