Learn More
Cayman Chemical MyrIstoylCoenzyme AsodIum 10mg
SDP
Catalog No. 502942122
Encompass_Preferred
A saturated acyl CoA; a substrate for N-myristoyltransferase during myristoylation; a substrate in the de novo synthesis of phosphatidylinositol
Catalog No. 50-294-2122 Supplier Cayman Chemical Supplier No. 3977710 MG
A saturated acyl CoA; a substrate for N-myristoyltransferase during myristoylation; a substrate in the de novo synthesis of phosphatidylinositol