Learn More
Cayman Chemical Myristoyl-L-carnitine (chloride), Purity: ≥98%, CAS: 173686-73-2, 10 mg
SDP
Catalog No. 502935050
Encompass_Preferred
A long-chain acylcarnitine; plasma levels are decreased in patients with chronic fatigue syndrome and increased in patients with end-stage renal disease
Catalog No. 50-293-5050 Supplier Cayman Chemical Supplier No. 2655910 MG
A long-chain acylcarnitine; plasma levels are decreased in patients with chronic fatigue syndrome and increased in patients with end-stage renal disease