Learn More
Cayman Chemical Myristoyl-L-carnitine (chloride), Purity: ≥98%, CAS: 173686-73-2, 50 mg
SDP
Catalog No. 502934502
Encompass_Preferred
A long-chain acylcarnitine; plasma levels are decreased in patients with chronic fatigue syndrome and increased in patients with end-stage renal disease
Catalog No. 50-293-4502 Supplier Cayman Chemical Supplier No. 2655950 MG
A long-chain acylcarnitine; plasma levels are decreased in patients with chronic fatigue syndrome and increased in patients with end-stage renal disease