Learn More
Medchemexpress LLC HY-133027 100mg Medchemexpress, Myristyl nicotinate CAS:273203-62-6 Purity:>98%
SDP
Catalog No. 501969848
Encompass_Preferred
Medchemexpress, HY-133027 100mg Myristyl nicotinate CAS:273203-62-6 Myristyl nicotinate (Tetradecyl nicotinate) is an ester prodrug and a lipophilic derivative of Nicotinic acid. Myristyl nicotinate is being developed for delivery of Nicotinic acid into the skin for prevention of actinic keratosis and its progression to skin cancer. Myristyl nicotinate shows to stimulate epidermal differentiation in photodamaged skin, increasing skin NAD content and strengthening the skin barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9848 Supplier Medchemexpress LLC Supplier No. HY133027100MG
Medchemexpress, HY-133027 100mg Myristyl nicotinate CAS:273203-62-6 Myristyl nicotinate (Tetradecyl nicotinate) is an ester prodrug and a lipophilic derivative of Nicotinic acid. Myristyl nicotinate is being developed for delivery of Nicotinic acid into the skin for prevention of actinic keratosis and its progression to skin cancer. Myristyl nicotinate shows to stimulate epidermal differentiation in photodamaged skin, increasing skin NAD content and strengthening the skin barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.