Learn More
Myron L Company Myron L D-1 Meter, Automatic Temperature Compensation, Rugged Construction, Less Than 1 lb
SDP
Catalog No. NC0996208
Encompass
The MYRON L COMPANY D-1 Meter is an advanced analog conductivity meter aimed at ensuring patient safety through precise and rapid testing of dialysate solutions in hemodialysis. Its key features include automatic temperature compensation and a user-friendly design with push-button operation. Designed for durability, its rugged construction makes it suitable for clinical environments, making it a reliable choice for healthcare professionals.
Catalog No. NC0996208 Supplier Myron L Company Supplier No. D1
The MYRON L COMPANY D-1 Meter is an advanced analog conductivity meter aimed at ensuring patient safety through precise and rapid testing of dialysate solutions in hemodialysis. Its key features include automatic temperature compensation and a user-friendly design with push-button operation. Designed for durability, its rugged construction makes it suitable for clinical environments, making it a reliable choice for healthcare professionals.