Learn More
Mitchell Instrument Company Inc MYRON L ULTRAMETER 4P II WATER
SDP
Catalog No. NC0855226
Encompass_Preferred
Measure Conductivity, TDS, Resistivity and Temperature with this Myron L Ultrameter all-in-one multiparameter meter. The Ultrameter II model delivers performance of +/- 1% of reading (not merely full scale).
Catalog No. NC0855226 Supplier Mitchell Instrument Company Inc Supplier No. MLJU4P
Measure Conductivity, TDS, Resistivity and Temperature with this Myron L Ultrameter all-in-one multiparameter meter. The Ultrameter II model delivers performance of +/- 1% of reading (not merely full scale).