Learn More
Mitchell Instrument Company Inc Myron L Ultrameter II 6PFCE Water Quality Meter
SDP
Catalog No. NC1704946
Encompass_Preferred
6PIIFCE Water Quality Meter. Measure pH, ORP (with free chlorine equivalency mode), Conductivity, TDS, Resistivity and Temperature with this Myron L Ultrameter all-in-one water quality meter. The Ultrameter II model delivers performance of ±1% of reading (not merely full scale). This high level of accuracy has been achieved through advanced four-electrode conductivity cell technology, a unique pH/ORP sensor and powerful microprocessor-based circuitry. Ideal for water quality applications in cooling towers, hydroponics, wastewater, reverse osmosis and much more.
Catalog No. NC1704946 Supplier Mitchell Instrument Company Inc Supplier No. MLJU6P
6PIIFCE Water Quality Meter. Measure pH, ORP (with free chlorine equivalency mode), Conductivity, TDS, Resistivity and Temperature with this Myron L Ultrameter all-in-one water quality meter. The Ultrameter II model delivers performance of ±1% of reading (not merely full scale). This high level of accuracy has been achieved through advanced four-electrode conductivity cell technology, a unique pH/ORP sensor and powerful microprocessor-based circuitry. Ideal for water quality applications in cooling towers, hydroponics, wastewater, reverse osmosis and much more.