Learn More
Cayman Chemical MyrTatCBD3trIfluoroACTAt 10mg
SDP
Catalog No. 502918187
Encompass_Preferred
An inhibitor of the Cav2.2-CRMP2 protein-protein interaction; inhibits the Cav2.2-CRMP2 interaction by ~81% at 10 µM and potassium-induced calcium influx in primary rat DRG neurons (IC50 = ~2.8 µM); inhibits calcium, but not sodium, currents in primary DRG neurons at 20 µM; intrathecal administration prevents carrageenan-induced decreases in paw withdrawal latencies in rats; decreases cue-induced reinstatement of cocaine-seeking behavior in rats
Catalog No. 50-291-8187 Supplier Cayman Chemical Supplier No. 3746510 MG
An inhibitor of the Cav2.2-CRMP2 protein-protein interaction; inhibits the Cav2.2-CRMP2 interaction by ~81% at 10 µM and potassium-induced calcium influx in primary rat DRG neurons (IC50 = ~2.8 µM); inhibits calcium, but not sodium, currents in primary DRG neurons at 20 µM; intrathecal administration prevents carrageenan-induced decreases in paw withdrawal latencies in rats; decreases cue-induced reinstatement of cocaine-seeking behavior in rats