Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Myrtenol, 97%, Thermo Scientific™

Catalog No. AC182581000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182581000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC182581000 Supplier Thermo Scientific Chemicals Supplier No. 182581000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19894-97-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00001341
InChI Key RXBQNMWIQKOSCS-IUCAKERBSA-N
Synonym --myrtenol, myrtenol,-, 1r---myrtenol, unii-s1b8392iqy, 1r-2-pinen-10-ol, 1r-6,6-dimethylbicyclo 3.1.1 hept-2-ene-2-methanol, 1r,5s-6,6-dimethylbicyclo 3.1.1 hept-2-en-2-yl methanol, 1s-+-myrtenol, --myrtenol,97
PubChem CID 88301
IUPAC Name [(1R,5S)-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl]methanol
SMILES CC1(C)[C@@H]2C[C@H]1C(CO)=CC2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.982g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4940 to 1.4990 (20°C, 589nm)
Quantity 100 g
Specific Rotation Condition −47° (20°C neat)
Specific Rotation -42° to -52° (20°C, 589nm) (neat)
Formula Weight 152.24
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material (-)-Myrtenol, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.