Learn More
Aviva Systems Biology Corp MYST2 Antibody - N-terminal region (ARP32842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYST2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MYST2. Alternate names: HBO1, HBOA, KAT7, MYST2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54791