Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mystaire™ CA Series Cyanoacrylate Fuming Chamber
GSA_VASDP

Catalog No. p-3975585 Shop All Mystaire Products
Change view
Click to view available options
Dimensions (L x W x H):
32 x 39 x 53 in.
32 x 39 x 65 in.
50 x 72 x 90 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
15-338-306 32 x 39 x 53 in.
15-338-307 32 x 39 x 65 in.
15-338-308 50 x 72 x 90 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 15-338-306 Supplier Mystaire™ Supplier No. CA3000
Only null left
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Safely fume evidence with consistent, reproducible results with the Mystaire™ CA Series Cyanoacrylate Fuming Chamber — a fully automated cyanoacrylate fingerprinting system.

Extensive Safety Features

  • Patented recirculatory ductless filtration
  • Door automatically locks during fuming
  • Seamless gasket ensures door is sealed
  • Custom filter for cyanoacrylate fumes
  • 500 cycles per cyanoacrylate filter
  • 50 cycles per prefilter
Precision Controls
  • Fully automated system
  • Accurate humidity control to 80%
  • Customizable fuming and filtering intervals
  • Display of options and parameters
  • Programmable internal circulation fan
  • High-temperature hotplate, 160°C
Superior Design and Construction
  • Powder-coated aluminum and steel frame
  • Safety glass door
  • Easy-to-clean interior
  • Sealed internal fluorescent lighting
  • Customizable shelving and interior features

CA-3000 and CA-6000 Models

  • Tabletop ductless units
  • Humidity control, heat source, internal circulation fan
  • Microprocessor control
  • Carbon filtering system captures hazardous fumes
  • All systems ship complete with filters
CA-9000 Model
  • High-volume, stand-alone fingerprinting chamber
  • Can be ductless or ducted to outside
  • Designed to U. S. government standards
  • Ideal for processing large amounts of evidence on a regular basis
  • Accommodates doors, tires, bicycles, tents and other large items

Order Info

For 110V 60Hz or 220V 50Hz.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Cyanoacralate Fuming Chamber
Coating Epoxy Powder
Filter Type Carbon
Material Epoxy Powder-coated Steel
Depth (English) 32 in.
Depth (English) Interior 25 in.
Depth (Metric) 813 mm
Depth (Metric) Interior 635 mm
Dimensions (D x W x H) 25 x 29.5 x 29 in.
Dimensions (L x W x H) 32 x 39 x 53 in.
Height (English) 53 in.
Height (English) Interior 36 in.
Height (Metric) 1346 mm
Height (Metric) Interior 737 mm
Hertz 50/60 Hz
Includes FE-2013 prefilter and MY-2818 main filter
Length (English) 32 in.
Length (Metric) 813 mm
Temperature Range (English) 320°F
Temperature Range (Metric) 160°C
Voltage 110/220 V
Weight (English) 265 lb.
Weight (Metric) 120 kg
Width (English) 39 in.
Width (English) Interior 29.5 in.
Width (Metric) 991 mm
Width (Metric) Interior 749 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.