Learn More
Mystaire™ Aura™ Main Filter
SDP
Catalog No. 15338190 Shop All Mystaire Products
Change view
Click to view available options
For Use With (Application):
Ammonia, Amines
Custom Blend
Formaldehyde
HEPA Particulates Application
Hydrocarbons
Mineral Acids
Multi-purpose
Particulate Air
Sulfur
For Use With (Equipment):
Aura 30, 54 Fume Hood
Aura 30, 54, 750 Fume Hood
Aura 42, 250, 550 Fume Hood
Material:
Gas Phase Carbon
HEPA
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) For Use With (Equipment) Material
15-338-190 Sulfur Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-181 Ammonia, Amines Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-207 Ammonia, Amines Aura 42, 250, 550 Fume Hood Gas Phase Carbon
15-338-122 Custom Blend Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-115 Custom Blend Aura 42, 250, 550 Fume Hood Gas Phase Carbon
15-338-118 Formaldehyde Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-176 Formaldehyde Aura 42, 250, 550 Fume Hood Gas Phase Carbon
15-338-113 Particulate Air Aura 42, 250, 550 Fume Hood HEPA
15-338-117 Hydrocarbons Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-110 Hydrocarbons Aura 42, 250, 550 Fume Hood Gas Phase Carbon
15-338-119 Mineral Acids Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-112 Mineral Acids Aura 42, 250, 550 Fume Hood Gas Phase Carbon
15-338-121 Multi-purpose Aura 30, 54, 750 Fume Hood Gas Phase Carbon
15-338-114 Multi-purpose Aura 42, 250, 550 Fume Hood Gas Phase Carbon
15-338-120 HEPA Particulates Application Aura 30, 54 Fume Hood HEPA
15-338-186 Sulfur Aura 42, 250, 550 Fume Hood Gas Phase Carbon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 15-338-190 Supplier Mystaire™ Supplier No. AU3809
Only null left
Add to Cart
Add to Cart
For Aura™ Series Ductless Fume Hoods. Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
For Use With (Equipment) Aura 30, 54, 750 Fume Hood
Product Type Filter
Material Gas Phase Carbon
For Use With (Application) Sulfur
WARNING: Cancer - www.P65Warnings.ca.gov