Learn More
Thomas Scientific MyTaq Extract-PCR Kit (500 reactions)
SDP
Catalog No. NC1682921
Encompass
Many DNA extraction methods can be laborious and time consuming or involve the use of hazardous chemicals. MyTaq™ Extract-PCR Kit offers a rapid, easy and safer alternative for the extraction and amplification of DNA from a variety of tissue types. MyTaq Extract-PCR Kit is particularly suited to solid tissues such as mouse tail or mouse ear. The DNA extractions are performed in a single-tube, without the need for multiple washing steps, greatly reducing the risk of sample loss and contamination.
Catalog No. NC1682921 Supplier Thomas Scientific Supplier No. C755G87
Many DNA extraction methods can be laborious and time consuming or involve the use of hazardous chemicals. MyTaq™ Extract-PCR Kit offers a rapid, easy and safer alternative for the extraction and amplification of DNA from a variety of tissue types. MyTaq Extract-PCR Kit is particularly suited to solid tissues such as mouse tail or mouse ear. The DNA extractions are performed in a single-tube, without the need for multiple washing steps, greatly reducing the risk of sample loss and contamination.