Learn More
Meridian Life Science MyTaq HS DNA Polymerase, 250 units
SDP
Catalog No. NC0995419
Encompass
A new generation of hot-start polymerase that delivers improved specificity, yield, speed and robustness when amplifying targets from any template.
Catalog No. NC0995419 Supplier Meridian Life Science Supplier No. BIO21111
A new generation of hot-start polymerase that delivers improved specificity, yield, speed and robustness when amplifying targets from any template.