Learn More
Thomas Scientific MyTaq HS DNA Polymerase (250 units)
SDP
Catalog No. NC1713216
Encompass
MyTaq HS DNA Polymerase is a new generation of antibody-mediated hot-start enzyme, engineered for highly specific and efficient amplification from even the most challenging templates. MyTaq HS remains inactive at room temperature allowing for convenient reaction set-up, thereby reducing non-specific amplification that can hinder PCR assays from the start. These properties make MyTaq HS DNA Polymerase the ideal choice for PCR assays containing complex and low copy number targets.
Catalog No. NC1713216 Supplier Thomas Scientific Supplier No. C755G78
MyTaq HS DNA Polymerase is a new generation of antibody-mediated hot-start enzyme, engineered for highly specific and efficient amplification from even the most challenging templates. MyTaq HS remains inactive at room temperature allowing for convenient reaction set-up, thereby reducing non-specific amplification that can hinder PCR assays from the start. These properties make MyTaq HS DNA Polymerase the ideal choice for PCR assays containing complex and low copy number targets.