Learn More
Meridian Life Science MYTAQ RED HS MIX
SDP
Catalog No. NC0321344
Encompass
MyTaq HS Red Mix is a ready-to-use 2x mix for fast, highly-specific hot-start PCR. MyTaq HS Red Mix is powered by antibody mediated hot-start and does not possess polymerase activity during the reaction set-up.
Catalog No. NC0321344 Supplier Meridian Life Science Supplier No. BIO25047
MyTaq HS Red Mix is a ready-to-use 2x mix for fast, highly-specific hot-start PCR. MyTaq HS Red Mix is powered by antibody mediated hot-start and does not possess polymerase activity during the reaction set-up.