Learn More
Alkali Scientific MyTemp 65L H2265-HC Lab Incuba
SDP
Catalog No. 503029552
Encompass_Preferred
The myTemp 65HC Incubator is designed to provide accurate and uniform temperature control from 0 to 60C The broad temperature range of the instrument expands the use of this incubator to both heating and cooling applications A large digital display allows for visualization of the chamber temperature and the remaining incubation time while an internal light is included for improved visualization of the chamber In addition a built in internal outlet can be used to provide power to small instruments such as shakers rockers etc With forced air technology and stainless steel inner construction uniform heat transfer is ensured throughout the internal chamber When set for sub-ambient temperatures the powerful compressor is activated quickly cooling the chamber to the desired temperature With an exterior height of 39 inches (100cm) the incubator is designed to be floor-standing yet is also compact enough for bench-use (Not recommended for use on benchtops taller than 32/81cm) The inst
Catalog No. 50-302-9552 Supplier Alkali Scientific Supplier No. RS7168
The myTemp 65HC Incubator is designed to provide accurate and uniform temperature control from 0 to 60C The broad temperature range of the instrument expands the use of this incubator to both heating and cooling applications A large digital display allows for visualization of the chamber temperature and the remaining incubation time while an internal light is included for improved visualization of the chamber In addition a built in internal outlet can be used to provide power to small instruments such as shakers rockers etc With forced air technology and stainless steel inner construction uniform heat transfer is ensured throughout the internal chamber When set for sub-ambient temperatures the powerful compressor is activated quickly cooling the chamber to the desired temperature With an exterior height of 39 inches (100cm) the incubator is designed to be floor-standing yet is also compact enough for bench-use (Not recommended for use on benchtops taller than 32/81cm) The inst