Learn More
Grainger Benchmark Scientific Mytemp 65L Incubator, 0°C to 60°C, Forced Air, 1 ea
Catalog No. 502786669
Encompass_Preferred
The myTemp 65HC Incubator provides accurate and uniform temperature control for a range of 0°C to 60°C, facilitating both heating and cooling applications. Featuring forced air technology and a stainless steel inner chamber, it ensures consistent heat transfer. The unit includes a digital display for monitoring temperature and incubation time, an internal light, and a built-in internal power outlet for auxiliary instruments.
  • Broad temperature range of 0°C to 60°C
  • Digital temperature display and control with 0.1°C increments
  • Forced air technology for uniform temperature distribution
  • Stainless steel interior construction with two adjustable height shelves
  • Built-in internal power outlet for operation of small instruments
  • Powerful compressor for sub-ambient temperature cooling
  • Compact, floor-standing design suitable for benchtop use
Catalog No. 50-278-6669 Supplier Grainger Supplier No. 198R84
The myTemp 65HC Incubator provides accurate and uniform temperature control for a range of 0°C to 60°C, facilitating both heating and cooling applications. Featuring forced air technology and a stainless steel inner chamber, it ensures consistent heat transfer. The unit includes a digital display for monitoring temperature and incubation time, an internal light, and a built-in internal power outlet for auxiliary instruments.
  • Broad temperature range of 0°C to 60°C
  • Digital temperature display and control with 0.1°C increments
  • Forced air technology for uniform temperature distribution
  • Stainless steel interior construction with two adjustable height shelves
  • Built-in internal power outlet for operation of small instruments
  • Powerful compressor for sub-ambient temperature cooling
  • Compact, floor-standing design suitable for benchtop use