Learn More
Thomas Scientific myTemp Mini CO2 Digital Incubator, 115V
SDP
Catalog No. 501457574
Encompass_Preferred
At only 13 inches wide, the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring CO2 gas control. Its small footprint makes the incubator ideal for use inside of biological safety cabinets, isolating sensitive cultures or for any laboratory simply trying to conserve bench space. With a thick layer of high efficiency insulation surrounding the chamber, the incubator is also ideal for transportation of cells. Once power has been disconnected, the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour*. In addition, each model includes a 12V vehicle power adapter for operating the instrument in a car, boat, etc. Both temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. This panel also features a "Cal" function, for easy in-lab calibration of the temperature or CO2. In addition, humidity (up to 80%) can be added to the chamber by inserting the include
Catalog No. 50-145-7574 Supplier Thomas Scientific Supplier No. 1186W48
At only 13 inches wide, the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring CO2 gas control. Its small footprint makes the incubator ideal for use inside of biological safety cabinets, isolating sensitive cultures or for any laboratory simply trying to conserve bench space. With a thick layer of high efficiency insulation surrounding the chamber, the incubator is also ideal for transportation of cells. Once power has been disconnected, the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour*. In addition, each model includes a 12V vehicle power adapter for operating the instrument in a car, boat, etc. Both temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. This panel also features a "Cal" function, for easy in-lab calibration of the temperature or CO2. In addition, humidity (up to 80%) can be added to the chamber by inserting the include