Learn More
Andwin Scientific MYTEMP MINI DIGI INCUBATOR EA
SDP
Catalog No. 50999402
Encompass_Preferred
The Benchmark MyTemp 115V Mini Digital Incubator allows digital display and control of heating and cooling and monitoring the chamber temperature. It features large internal chambers that can store 2L flask or bottles. It is sold per piece.
Catalog No. 50-999-402 Supplier Andwin Scientific Supplier No. H2200HC
The Benchmark MyTemp 115V Mini Digital Incubator allows digital display and control of heating and cooling and monitoring the chamber temperature. It features large internal chambers that can store 2L flask or bottles. It is sold per piece.