Learn More
Research Products International Corp MyTemp Mini Digital Incubator, Heat Only Model
SDP
Catalog No. 501977101
Encompass
With digital temperature control, the MyTemp incubators provide convenient set and walk away operation, eliminating the need for external thermometers and repetitive fine tuning of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up (or cool down, depending on model) while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Despite a modest footprint of 14.5 x 13", the MyTemp incubators feature large internal chambers, capable of storing flasks and bottles up to 2 liter. In addition, the incubators include two adjustable/removeable shelves for increased capacity. Units feature Peltier chip with temperature sensor.
2 Year Warranty.
Catalog No. 50-197-7101 Supplier Research Products International Corp Supplier No. 112150
With digital temperature control, the MyTemp incubators provide convenient set and walk away operation, eliminating the need for external thermometers and repetitive fine tuning of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up (or cool down, depending on model) while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Despite a modest footprint of 14.5 x 13", the MyTemp incubators feature large internal chambers, capable of storing flasks and bottles up to 2 liter. In addition, the incubators include two adjustable/removeable shelves for increased capacity. Units feature Peltier chip with temperature sensor.
2 Year Warranty.