Learn More
Labrepco Inc MyTemp Mini Digital Incubator, 115V, Heat Only
SDP
Catalog No. NC0936027
Encompass_Preferred
MyTemp Mini Digital Incubator, 115V (Heat Only) With digital temperature control, the MyTemp incubator provides convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Specifications: Temperature Range: ambient +1C to +60C Temperature Accuracy: 0.5C (at +37C) Temperature Uniformity: +/- 1C (at 37C) Temperature Increments: +/- 1C Exterior Dimensions: 18.7"H x 14.5"W x 13.2"D Interior Dimensions: 12.8"H x 9.3"W x 10.3"D Capacity: 20L Weight: 15 lbs Electrical: 115V, 50-60Hz Features: Digital temperature control Personal sized, 13 x 14.5" footprint Accepts bottles and flasks up to 12L Optional 3D mini shaker available for sample agitation
Catalog No. NC0936027 Supplier Labrepco Inc Supplier No. H2200H
MyTemp Mini Digital Incubator, 115V (Heat Only) With digital temperature control, the MyTemp incubator provides convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Specifications: Temperature Range: ambient +1C to +60C Temperature Accuracy: 0.5C (at +37C) Temperature Uniformity: +/- 1C (at 37C) Temperature Increments: +/- 1C Exterior Dimensions: 18.7"H x 14.5"W x 13.2"D Interior Dimensions: 12.8"H x 9.3"W x 10.3"D Capacity: 20L Weight: 15 lbs Electrical: 115V, 50-60Hz Features: Digital temperature control Personal sized, 13 x 14.5" footprint Accepts bottles and flasks up to 12L Optional 3D mini shaker available for sample agitation