Learn More
Thomas Scientific Mini Nutating Rocker, 8 x 6" platform, 115V
SDP
Catalog No. 501460140
Encompass_Preferred
Digital temperature control Personal sized, 13 x 14.5" footprint (20L) Accepts bottles and flasks up to 2L Two models: Heat Only or Heat/Cool Internal outlet, for installation of shaker, rockers, etc. With digital temperature control, the MyTemp incubators provide convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up (or cool down, -HC model only) while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Despite a modest footprint of 14.5 x 13″, the MyTemp incubators feature large internal chambers, capable of storing flasks and bottles up to 2 liter. In addition, the incubators include two adjustable shelves and an internal outlet for powering small shakers, rockers and other instrumentation. A mini
Catalog No. 50-146-0140 Supplier Thomas Scientific Supplier No. 1159P22
Digital temperature control Personal sized, 13 x 14.5" footprint (20L) Accepts bottles and flasks up to 2L Two models: Heat Only or Heat/Cool Internal outlet, for installation of shaker, rockers, etc. With digital temperature control, the MyTemp incubators provide convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up (or cool down, -HC model only) while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Despite a modest footprint of 14.5 x 13″, the MyTemp incubators feature large internal chambers, capable of storing flasks and bottles up to 2 liter. In addition, the incubators include two adjustable shelves and an internal outlet for powering small shakers, rockers and other instrumentation. A mini