Learn More
Sigma Aldrich Fine Chemicals Biosciences MyTempTM Mini CO2 Digital1EA
Catalog No. 502908714
Encompass
At only 13 inches wide the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring CO2 gas control. Its small footprint makes the incubator ideal for use inside of biological safety cabinets isolating sensitive cultures or for any laboratory simply trying to conserve bench space. With a thick layer of high efficiency insulation surrounding the chamber the incubator is also ideal for transportation of cells. Once power has been disconnected the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour*. In addition each model includes a 12V vehicle power adapter for operating the instrument in a car boat etc.Both temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. This panel also features a Cal function for easy in-lab calibration of the temperature or CO2. In addition humidity (up to 80%) can be added to the chamber by inserting the included (stainless s
Catalog No. 50-290-8714 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7425321EA
At only 13 inches wide the myTemp Mini CO2 is the smallest and most economical incubator for applications requiring CO2 gas control. Its small footprint makes the incubator ideal for use inside of biological safety cabinets isolating sensitive cultures or for any laboratory simply trying to conserve bench space. With a thick layer of high efficiency insulation surrounding the chamber the incubator is also ideal for transportation of cells. Once power has been disconnected the incubator maintains temperature levels within 10% of the set parameter for up to 1 hour*. In addition each model includes a 12V vehicle power adapter for operating the instrument in a car boat etc.Both temperature and CO2 percentage are digitally adjustable and displayed in real time on the large LED control panel. This panel also features a Cal function for easy in-lab calibration of the temperature or CO2. In addition humidity (up to 80%) can be added to the chamber by inserting the included (stainless s