Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mytilus edulis foot protein-1

Catalog No. 7677001ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-770-01ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-770-01ML Supplier Sigma Aldrich Supplier No. 7677001ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A solution of MEFP-1 that is specially developed for biological applications. The protein can be used to coat surfaces in order to facilitate immobilization of cells and tissue. The solution is sterile by autoclaving and has an endotoxin level of = 1 EU/ml. The product is cell culture tested (cytotoxicity and cell adhesion assay). MEFP-1 can be successfully used for unspecific immobilization of cells and tissue to essentially any surface. It is an especially useful tool when the surface chemistry results in poor adhesion of cells or for cells that are difficult to attach. In comparison to binding strategies based on electrostatic interactions (positively charged surfaces) or covalent immobilization (EDC/NHS or glutaraldehyde) attachment using MEFP-1 is insensitive to ionic strength and pH and will not result in undesired chemical modification, such as cross-linking, of cell surface components. The protein coating is a useful tool in techniques such as in situ hybridization, immunohistochemistry, tissue engineering, drug delivery, ELISA, microfluidics, microinjection and various microscopy techniques.

Specifications

Density 1.008 g/mL (at 25°C)
Boiling Point 100°C to 103°C (760 mmHg)
Refractive Index n20/D 1.335 (literature)
Quantity 1 mL
Synonym MAPcell
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.