Learn More
Alkali Scientific Alkali Scientific™ myVolt™ Mini power supply, 115V
SDP
Catalog No. 503029643
Encompass_Preferred
The Alkali Scientific myVolt Mini ES2100 is a compact electrophoresis power supply ideal for basic, constant voltage applications. Users can select constant 100V or 200V output, and the current can reach a maximum of 200mA. Two sets of output jacks are available to connect up to 2 gel rigs and these small power supplies can securely be stacked for use with additional gel rigs. Compact and simple Choose constant 100V or 200V output Two pairs of output terminals
Catalog No. 50-302-9643 Supplier Alkali Scientific Supplier No. E2100
The Alkali Scientific myVolt Mini ES2100 is a compact electrophoresis power supply ideal for basic, constant voltage applications. Users can select constant 100V or 200V output, and the current can reach a maximum of 200mA. Two sets of output jacks are available to connect up to 2 gel rigs and these small power supplies can securely be stacked for use with additional gel rigs. Compact and simple Choose constant 100V or 200V output Two pairs of output terminals