Learn More
AdipoGen Myxochelin A (100 µg)
SDP
Supplier:  AdipoGen AGCN20470C100
Encompass_Preferred
Chemical. CAS: 120243-02-9. Formula: C20H24N2O7. MW: 404.4. Synthetic. Originally isolated from Pyxidicoccus fallax HKI 727. Potent inhibitor of human 5-lipoxygenase (5-LO). This enzyme catalyzes two initial steps in the conversion of arachidonic acid into leukotrienes, well known mediators of inflammatory and allergic reactions. Iron-chelating compound. Anticancer antibiotic. Shown to inhibit tumor cell invasion in vitro. Antibacterial compound. Antioxidant with free radical scavenging activities.
Catalog No. 50-205-3651