Learn More
Sigma Aldrich Fine Chemicals Biosciences Myxothiazol, 76706-55-3, MFCD00043397, 1 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T55801MG
Encompass
Empirical Formula (Hill Notation): C25H33N3O3S2, Molecular Weight: 487.68, from Myxococcus fulvus Mx f85, ≥98% (HPLC), storage condition: protect from light.
Catalog No. NC0862027