Learn More
Medchemexpress LLC Myxothiazol | 76706-55-3 | >98.0% | 487.68 g·mol⁻¹ | C25H33N3O3S2 | 5 MG
SDP
Catalog No. 5000263747
Encompass_Preferred
Myxothiazol is an antifungal antibiotic that selectively inhibits the mitochondrial electron transport chain complex III (bc1 complex). Supplied as a research-grade small-molecule solid, it is used in biochemical and cellular studies to probe mitochondrial respiration, assess respiratory chain inhibition, and study antifungal activity. CAS: 76706-55-3; formula: C25H33N3O3S2; molar mass: 487.68 g·mol⁻¹.
  • Inhibits mitochondrial complex III (bc1 complex) for targeted mechanistic studies.
  • Useful for investigating mitochondrial respiration and cellular bioenergetics.
  • Applicable to in vitro biochemical assays and fungal growth inhibition studies.
  • Available in small research pack sizes for precise dosing and formulation.
  • Well characterized with CAS, molecular formula, and molecular weight identifiers.
Catalog No. 50-002-63747 Supplier Medchemexpress LLC Supplier No. HY1121775MG
Myxothiazol is an antifungal antibiotic that selectively inhibits the mitochondrial electron transport chain complex III (bc1 complex). Supplied as a research-grade small-molecule solid, it is used in biochemical and cellular studies to probe mitochondrial respiration, assess respiratory chain inhibition, and study antifungal activity. CAS: 76706-55-3; formula: C25H33N3O3S2; molar mass: 487.68 g·mol⁻¹.
  • Inhibits mitochondrial complex III (bc1 complex) for targeted mechanistic studies.
  • Useful for investigating mitochondrial respiration and cellular bioenergetics.
  • Applicable to in vitro biochemical assays and fungal growth inhibition studies.
  • Available in small research pack sizes for precise dosing and formulation.
  • Well characterized with CAS, molecular formula, and molecular weight identifiers.