Learn More
Medchemexpress LLC Myxothiazol | 76706-55-3 | 99.5% | 487.68 | 1 MG
SDP
Catalog No. 5000437083
Encompass_Preferred
Myxothiazol is an antifungal antibiotic that acts as a mitochondrial electron transport chain complex III (bc1 complex) inhibitor. It inhibits the growth of various yeasts and fungi at concentrations between 0.01 and 3 μg/ml. Myxothiazol binds to the ubiquinol oxidation site Qo of complex III, blocking electron transfer from ubiquinol to cytochrome b and thereby inhibiting complex III activity.
  • Antifungal antibiotic
  • Mitochondrial electron transport chain complex III (bc1 complex) inhibitor
  • Inhibits growth of many yeasts and fungi
  • For research use only
Catalog No. 50-004-37083 Supplier Medchemexpress LLC Supplier No. HY1121771MG
Myxothiazol is an antifungal antibiotic that acts as a mitochondrial electron transport chain complex III (bc1 complex) inhibitor. It inhibits the growth of various yeasts and fungi at concentrations between 0.01 and 3 μg/ml. Myxothiazol binds to the ubiquinol oxidation site Qo of complex III, blocking electron transfer from ubiquinol to cytochrome b and thereby inhibiting complex III activity.
  • Antifungal antibiotic
  • Mitochondrial electron transport chain complex III (bc1 complex) inhibitor
  • Inhibits growth of many yeasts and fungi
  • For research use only